
2-(Chloromethyl)-3-methylpyridine hydrochloride 95%
Catalog # APOSOR480862-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.059 g/mol
Melting Pt:159 °C
:GHS07
Storage Temperature:Ambient
CAS Number:4370-22-3
Formula:C₇H₈ClN·ClH
MDL Number:MFCD08669778