Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl 5-chloro-4-nitro-thiophene-2-carboxylate 95%
Catalog # APOSOR480790-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5-chloro-4-nitro-thiophene-2-carboxylate 95%
Catalog # APOSOR480790-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :221.618 g/mol
Storage Temperature:Ambient
CAS Number:57800-76-7
Formula:C₆H₄ClNO₄S
MDL Number:MFCD08275718