Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
2-Chloro-1-methyl-4-nitro-1H-imidazole
2-Chloro-1-methyl-4-nitro-1H-imidazole 95%
2-Chloro-1-methyl-4-nitro-1H-imidazole 95%
2-Chloro-1-methyl-4-nitro-1H-imidazole 95%
APOSOR480731-5G
:
Apollo Scientific
:
2-Chloro-1-methyl-4-nitro-1H-imidazole 95%
APOSOR480731-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
161.546 g/mol
:
Ambient
:
63634-21-9
:
C₄H₄ClN₃O₂
:
MFCD01721662