1-Pyridazin-4-ylcyclopropanecarboxylic acid
Catalog # APOSOR480110-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:164.161 g/mol
Storage Temperature:Ambient
CAS Number:2090420-67-8
Formula:C₈H₈N₂O₂
MDL Number:MFCD30496513