1-(3-Chloro-2-pyridyl)cyclopropanecarboxylic acid
Catalog # APOSOR480109-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:197.618 g/mol
Storage Temperature:Ambient
CAS Number:1427020-55-0
Formula:C₉H₈ClNO₂
MDL Number:MFCD23134934