1-Pyridazin-4-ylcyclopropanecarbonitrile
Catalog # APOSOR480105-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:145.161 g/mol
Storage Temperature:Ambient
Formula:C₈H₇N₃
MDL Number:MFCD30496585