1-(1-Methylimidazol-2-yl)cyclopropanamine hydrochloride
Catalog # APOSOR480099-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:173.643 g/mol
Storage Temperature:Ambient
Formula:C₇H₁₁N₃·ClH
MDL Number:MFCD30496559