Order Entry
Czech Republic
ContactUsLinkComponent
1-(1-Methylimidazol-4-yl)cyclopropanamine hydrochloride
1-(1-Methylimidazol-4-yl)cyclopropanamine hydrochloride
Catalog # APOSOR480098-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(1-Methylimidazol-4-yl)cyclopropanamine hydrochloride
Catalog # APOSOR480098-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.643 g/mol
Storage Temperature:Ambient
CAS Number:1159737-42-4
Formula:C₇H₁₁N₃·ClH
MDL Number:MFCD30496556