1-(1-Methylpyrrol-3-yl)cyclopropanamine hydrochloride
Catalog # APOSOR480095-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:172.655 g/mol
Storage Temperature:Ambient
Formula:C₈H₁₂N₂·ClH
MDL Number:MFCD30496548