Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 5-amino-4-chloro-1-sec-butyl-pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-sec-butyl-pyrazole-3-carboxylate
Catalog # APOSOR480014-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-amino-4-chloro-1-sec-butyl-pyrazole-3-carboxylate
Catalog # APOSOR480014-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :245.706 g/mol
Storage Temperature:Ambient
CAS Number:1427024-19-8
Formula:C₁₀H₁₆ClN₃O₂
MDL Number:MFCD23135180