Ethyl 2-[1-(benzylamino)ethyl]benzoate hydrochloride
Catalog # APOSOR475121-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:319.826 g/mol
Storage Temperature:Ambient
Formula:C₁₈H₂₁NO₂·ClH
MDL Number:MFCD31695680