N-[(2-Bromophenyl)methyl]-1-phenyl-ethanamine hydrochloride
Catalog # APOSOR475115-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:326.659 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₆BrN·ClH
MDL Number:MFCD31695664