
1-(4-Bromophenyl)-N-methylethanamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:799293-97-3
:
Formula:C₉H₁₂BrN·ClH
MDL Number:MFCD31695722
:250.563 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together