
1-(3-Bromophenyl)-N-methylethanamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2197556-55-9
:
Formula:C₉H₁₂BrN·ClH
MDL Number:MFCD30609564
:250.563 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together