Order Entry
Czech Republic
ContactUsLinkComponent
(1S)-N-Benzyl-1-(3-bromophenyl)ethanamine hydrochloride
(1S)-N-Benzyl-1-(3-bromophenyl)ethanamine hydrochloride
Catalog # APOSOR475033-1G
Supplier:  Apollo Scientific
CAS Number:  
(1S)-N-Benzyl-1-(3-bromophenyl)ethanamine hydrochloride
Catalog # APOSOR475033-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :326.659 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₆BrN·ClH
MDL Number:MFCD31695710