Ethyl 2-[(1S)-1-(3-bromoanilino)ethyl]benzoate hydrochloride
Catalog # APOSOR475022-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:384.695 g/mol
Storage Temperature:Ambient
Formula:C₁₇H₁₈BrNO₂·ClH
MDL Number:MFCD31695681