Order Entry
Czech Republic
ContactUsLinkComponent
5-(2-Hydroxyethyl)nicotinonitrile 95%
5-(2-Hydroxyethyl)nicotinonitrile 95%
Catalog # APOSOR471730-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(2-Hydroxyethyl)nicotinonitrile 95%
Catalog # APOSOR471730-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :148.162 g/mol
Storage Temperature:Ambient
CAS Number:1694868-89-7
Formula:C₈H₈N₂O
MDL Number:MFCD31695766