Order Entry
Czech Republic
ContactUsLinkComponent
(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine 98%
(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine 98%
Catalog # APOSOR471649-5G
Supplier:  Apollo Scientific
CAS Number:  
(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine 98%
Catalog # APOSOR471649-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :273.349 g/mol
Storage Temperature:Ambient
CAS Number:608141-42-0
Formula:C₁₂H₁₉NO₄S
MDL Number:MFCD22677268