Order Entry
Czech Republic
ContactUsLinkComponent
2-(5-Ethyl-1,3,4-oxadiazol-2-yl)ethylamine
2-(5-Ethyl-1,3,4-oxadiazol-2-yl)ethylamine
Catalog # APOSOR471579-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(5-Ethyl-1,3,4-oxadiazol-2-yl)ethylamine
Catalog # APOSOR471579-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :141.171 g/mol
Storage Temperature:Ambient
CAS Number:944898-31-1
Formula:C₆H₁₁N₃O
MDL Number:MFCD09883756