Order Entry
Czech Republic
ContactUsLinkComponent
2-(6-Bromo-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
2-(6-Bromo-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Catalog # APOSOR471524-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(6-Bromo-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Catalog # APOSOR471524-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :312.912 g/mol
Storage Temperature:Ambient
CAS Number:1816951-75-3
Formula:C₁₀H₁₀BBrN₂O₄
MDL Number:MFCD11215211