Order Entry
Czech Republic
ContactUsLinkComponent
2-(2-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
2-(2-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Catalog # APOSOR471519-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Catalog # APOSOR471519-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :249.028 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₂BNO₅
MDL Number:MFCD29049233