Ethyl 2-[trans-4-[(Ethylamino)methyl]cyclohexyl]acetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:669080-89-1
:
Formula:C₁₃H₂₅NO₂
MDL Number:MFCD19105333
:227.343 g/mol
Storage Temperature:Ambient
Frequently Bought Together