Methyl cis-1-Boc-4-aminopiperidine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1620128-54-2
:
Formula:C₁₂H₂₂N₂O₄
MDL Number:MFCD21332897
:258.314 g/mol
Storage Temperature:Ambient
Frequently Bought Together