
Methyl cis-3-Aminocyclopentanecarboxylate hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:222530-29-2
:
Formula:C₇H₁₃NO₂·ClH
MDL Number:MFCD27922569
:179.645 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together