3-[(4-Chlorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Catalog # APOSOR471408-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:337.755 g/mol
Storage Temperature:Ambient
CAS Number:2006277-13-8
Formula:C₁₆H₁₆ClNO₅
MDL Number:MFCD28977382