Methyl 3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Catalog # APOSOR471403-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:312.116 g/mol
Storage Temperature:Ambient
CAS Number:2006277-99-0
Formula:C₁₂H₁₀BrNO₄
MDL Number:MFCD28977387