2-(Chloromethyl)-6,7-dimethoxyquinazoline
Catalog # APOSOR471377-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.67 g/mol
Storage Temperature:Ambient
CAS Number:2006276-97-5
Formula:C₁₁H₁₁ClN₂O₂
MDL Number:MFCD28977347