2-(3-Chloropropyl)-6,7-dimethoxyquinazoline
Catalog # APOSOR471375-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.723 g/mol
Storage Temperature:Ambient
CAS Number:2006277-49-0
Formula:C₁₃H₁₅ClN₂O₂
MDL Number:MFCD28977349