6-Bromo-2-(hydroxymethyl)-4-iodobenzimidazole
Catalog # APOSOR471356-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:352.955 g/mol
Storage Temperature:Ambient
CAS Number:1935040-02-0
Formula:C₈H₆BrIN₂O
MDL Number:MFCD28977367