4-Bromo-2-(hydroxymethyl)-6-methylbenzimidazole
Catalog # APOSOR471354-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:241.085 g/mol
Storage Temperature:Ambient
CAS Number:1352519-72-2
Formula:C₉H₉BrN₂O
MDL Number:MFCD28977369