4-Bromo-2-(hydroxymethyl)benzimidazole
Catalog # APOSOR471353-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.058 g/mol
Storage Temperature:Ambient
CAS Number:1248566-21-3
Formula:C₈H₇BrN₂O
MDL Number:MFCD17267703