Order Entry
Czech Republic
ContactUsLinkComponent
3,7-Dibromo-2,6-dihydroxynaphthalene
3,7-Dibromo-2,6-dihydroxynaphthalene
Catalog # APOSOR471332-1G
Supplier:  Apollo Scientific
CAS Number:  
3,7-Dibromo-2,6-dihydroxynaphthalene
Catalog # APOSOR471332-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :317.961 g/mol
Storage Temperature:Ambient
CAS Number:1227743-83-0
Formula:C₁₀H₆Br₂O₂
MDL Number:MFCD28954613