2-Chloro-3,6-dimethylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:72093-14-2
:
Formula:C₇H₈ClN
MDL Number:MFCD12400985
:141.598 g/mol
Boiling Pt:45 °C (80 mm Hg)
Storage Temperature:Refrigerator
Frequently Bought Together