Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 4-hydroxy-1-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate
Methyl 4-hydroxy-1-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate
Catalog # APOSOR471323-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-hydroxy-1-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate
Catalog # APOSOR471323-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.161 g/mol
Storage Temperature:Ambient
CAS Number:916226-83-0
Formula:C₈H₉NO₄
MDL Number:MFCD27986809