1-(2,4-Dimethoxyphenyl)cyclopropanecarboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1314763-76-2
:
Formula:C₁₂H₁₄O₄
MDL Number:MFCD19692943
:222.237 g/mol
Storage Temperature:Ambient
Frequently Bought Together