Order Entry
Czech Republic
ContactUsLinkComponent
6-(2-Chlorophenyl)morpholin-3-one
6-(2-Chlorophenyl)morpholin-3-one
Catalog # APOSOR471183-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(2-Chlorophenyl)morpholin-3-one
Catalog # APOSOR471183-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211.645 g/mol
Storage Temperature:Ambient
CAS Number:951626-19-0
Formula:C₁₀H₁₀ClNO₂
MDL Number:MFCD09831998