Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dimethoxy-3,5-dinitropyridine
2,6-Dimethoxy-3,5-dinitropyridine
2,6-Dimethoxy-3,5-dinitropyridine
APOSOR471159-1G
:
Apollo Scientific
:
2,6-Dimethoxy-3,5-dinitropyridine
APOSOR471159-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
229.147 g/mol
:
Ambient
:
18677-42-4
:
C₇H₇N₃O₆
:
MFCD00955977