Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2-Amino-3-pyridyl)(morpholino)methanone
(2-Amino-3-pyridyl)(morpholino)methanone
(2-Amino-3-pyridyl)(morpholino)methanone
APOSOR471151-1G
:
Apollo Scientific
:
(2-Amino-3-pyridyl)(morpholino)methanone
APOSOR471151-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
207.229 g/mol
:
Ambient
:
1248775-87-2
:
C₁₀H₁₃N₃O₂
:
MFCD16678947