3-Bromo-2-chloro-5-(hydroxymethyl)pyridine
Catalog # APOSOR471126-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:222.467 g/mol
Storage Temperature:Ambient
CAS Number:904745-59-1
Formula:C₆H₅BrClNO
MDL Number:MFCD15526782