Order Entry
Czech Republic
ContactUsLinkComponent
7-Bromo-2-chloro-5H-pyrrolo[3,2-d]pyrimidine
7-Bromo-2-chloro-5H-pyrrolo[3,2-d]pyrimidine
Catalog # APOSOR471125-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Bromo-2-chloro-5H-pyrrolo[3,2-d]pyrimidine
Catalog # APOSOR471125-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.465 g/mol
Storage Temperature:Ambient
CAS Number:1429882-36-9
Formula:C₆H₃BrClN₃
MDL Number:MFCD22398085