4-Amino-4'-hydroxybenzophenone
Catalog # APOSOR471121-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:213.232 g/mol
Storage Temperature:Ambient
CAS Number:14963-34-9
Formula:C₁₃H₁₁NO₂
MDL Number:MFCD02648435