N-[3-(Dimethylamino)propyl]acetamide
Catalog # APOSOR471086-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:144.215 g/mol
Storage Temperature:Ambient
CAS Number:3197-19-1
Formula:C₇H₁₆N₂O
MDL Number:MFCD01195313