Methyl 3-(1-Hydroxyethyl)benzoate
Catalog # APOSOR471065-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.2 g/mol
Storage Temperature:Ambient
CAS Number:87808-11-5
Formula:C₁₀H₁₂O₃
MDL Number:MFCD28122816