Order Entry
Czech Republic
ContactUsLinkComponent
N-[3-(2,2-Diethoxyethoxy)benzyl]-2,2-diethoxyethylamine
N-[3-(2,2-Diethoxyethoxy)benzyl]-2,2-diethoxyethylamine
Catalog # APOSOR471047-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[3-(2,2-Diethoxyethoxy)benzyl]-2,2-diethoxyethylamine
Catalog # APOSOR471047-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :355.469 g/mol
Storage Temperature:Ambient
CAS Number:2006277-32-1
Formula:C₁₉H₃₃NO₅
MDL Number:MFCD28101620