Order Entry
Czech Republic
ContactUsLinkComponent
3-(Bromomethyl)piperidine hydrochloride
3-(Bromomethyl)piperidine hydrochloride
Catalog # APOSOR471031-5G
Supplier:  Apollo Scientific
CAS Number:  
3-(Bromomethyl)piperidine hydrochloride
Catalog # APOSOR471031-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :214.531 g/mol
Storage Temperature:Ambient
CAS Number:1353976-07-4
Formula:C₆H₁₂BrN·ClH
MDL Number:MFCD21098325