5-[Hydroxy(phenyl)methyl]dihydrofuran-2(3H)-one
Catalog # APOSOR471026-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:192.211 g/mol
Storage Temperature:Ambient
CAS Number:92017-03-3
Formula:C₁₁H₁₂O₃
MDL Number:MFCD28053835