Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 6-(Isopropylamino)pyridazine-3-carboxylate
Ethyl 6-(Isopropylamino)pyridazine-3-carboxylate
Catalog # APOSOR470969-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 6-(Isopropylamino)pyridazine-3-carboxylate
Catalog # APOSOR470969-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :209.245 g/mol
Storage Temperature:Ambient
CAS Number:1823359-03-0
Formula:C₁₀H₁₅N₃O₂
MDL Number:MFCD28014559