3-Hydroxy-5-(methylsulphonamido)benzoic acid
Catalog # APOSOR470958-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:231.226 g/mol
Storage Temperature:Ambient
CAS Number:1243364-91-1
Formula:C₈H₉NO₅S
MDL Number:MFCD16999874