Methyl 5,5-dimethyl-2-oxocyclohexanecarboxylate
Catalog # APOSOR470921-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.232 g/mol
Storage Temperature:Ambient
CAS Number:50388-51-7
Formula:C₁₀H₁₆O₃
MDL Number:MFCD09869052