Order Entry
Czech Republic
ContactUsLinkComponent
5-Aminothiazolo[4,5-d]pyrimidin-2(3H)-one
5-Aminothiazolo[4,5-d]pyrimidin-2(3H)-one
Catalog # APOSOR470920-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Aminothiazolo[4,5-d]pyrimidin-2(3H)-one
Catalog # APOSOR470920-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.176 g/mol
Storage Temperature:Ambient
CAS Number:848691-22-5
Formula:C₅H₄N₄OS
MDL Number:MFCD17017931